ENA300791629 Enamine

4-(2-aminoethyl)benzene-1,3-diol hydrochloride

  • MDL number MFCD00798944
  • Cas Number : 1432-22-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65185

Safety Information

msds https://www.enaminestore.com/catalog/EN300-65185
Availability details for
Quantity
  • Availability
  • Add to Cart