ENA300791785 Enamine

4-(benzylsulfanyl)-1-phenyl-1H-imidazole-5-carbaldehyde

  • MDL number MFCD16990645
  • Cas Number : 1251922-54-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-67194
Availability details for
Quantity
  • Availability
  • Add to Cart