ENA300791790 Enamine

N,2-dimethyl-6-oxo-1,6-dihydropyrimidine-5-carboxamide

  • MDL number MFCD16990650
  • Cas Number : 1251925-07-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-67268

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67268
Availability details for
Quantity
  • Availability
  • Add to Cart