ENA300791791 Enamine

1-[(4-aminophenyl)sulfanyl]-N,N-dimethylformamide hydrochloride

  • MDL number MFCD16990651
  • Cas Number : 1251925-33-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67285

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67285
Availability details for
Quantity
  • Availability
  • Add to Cart