ENA300791793 Enamine

3-(aminomethyl)-N-tert-butylbenzamide hydrochloride

  • MDL number MFCD16990653
  • Cas Number : 1251923-41-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67327

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67327
Availability details for
Quantity
  • Availability
  • Add to Cart