ENA300791794 Enamine

5-methylpyrazolo[1,5-a]pyrimidine-3-carboxylic acid

  • MDL number MFCD16990654
  • Cas Number : 62908-85-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67334

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67334
Availability details for
Quantity
  • Availability
  • Add to Cart