ENA300791795 Enamine

2-[1-(2,2,2-trifluoroethyl)-1H-imidazol-2-yl]acetonitrile

  • MDL number MFCD16990655
  • Cas Number : 1251923-15-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67343

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67343
Availability details for
Quantity
  • Availability
  • Add to Cart