ENA300791802 Enamine

1-[(1-phenyl-1H-1,2,3,4-tetrazol-5-yl)methyl]piperazine dihydrochloride

  • MDL number MFCD16661038
  • Cas Number : 1251924-70-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 208...210
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67406

Safety Information

msds https://www.enaminestore.com/catalog/EN300-67406
Availability details for
Quantity
  • Availability
  • Add to Cart