ENA300791807 Enamine

2-(3-methylbutyl)cyclopentan-1-amine hydrochloride

  • MDL number MFCD16990667
  • Cas Number : 1251925-20-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67436

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67436
Availability details for
Quantity
  • Availability
  • Add to Cart