ENA300791809 Enamine

3-(2-fluorophenyl)-1H-1,2,4-triazol-5-amine hydrochloride

  • MDL number MFCD16990670
  • Cas Number : 1251922-61-8
  • Molecular Weight : 214.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-67507

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67507
Availability details for
Quantity
  • Availability
  • Add to Cart