ENA300791811 Enamine

3-[(4-amino-3-methylphenoxy)methyl]benzonitrile hydrochloride

  • MDL number MFCD16990673
  • Cas Number : 1251925-23-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67579

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67579
Availability details for
Quantity
  • Availability
  • Add to Cart