ENA300791815 Enamine

5-chloro-8-methoxy-1,2,3,4-tetrahydroquinoline hydrochloride

  • MDL number MFCD16990679
  • Cas Number : 1251925-38-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-67671

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67671
Availability details for
Quantity
  • Availability
  • Add to Cart