ENA300791817 Enamine

4-ethoxy-2,3-dihydro-1H-inden-1-amine hydrochloride

  • MDL number MFCD16990681
  • Cas Number : 1251923-48-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 227...229
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68163

Safety Information

msds https://www.enaminestore.com/catalog/EN300-68163
Availability details for
Quantity
  • Availability
  • Add to Cart