ENA300791819 Enamine

2-hydroxy-N-(3-nitrophenyl)acetamide

  • MDL number MFCD16990683
  • Cas Number : 859980-47-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68192

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68192
Availability details for
Quantity
  • Availability
  • Add to Cart