ENA300791820 Enamine

7-chlorofuro[2,3-d]pyridazine

  • MDL number MFCD16990684
  • Cas Number : 14757-89-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117...119
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68209

Safety Information

msds https://www.enaminestore.com/catalog/EN300-68209
Availability details for
Quantity
  • Availability
  • Add to Cart