ENA300791824 Enamine

3-(quinolin-2-yl)-1H-1,2,4-triazol-5-amine

  • MDL number MFCD16990688
  • Cas Number : 1018458-44-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-68239

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68239
Availability details for
Quantity
  • Availability
  • Add to Cart