ENA300791825 Enamine

2-amino-2-[2-(trifluoromethyl)phenyl]acetonitrile hydrochloride

  • MDL number MFCD16990689
  • Cas Number : 127952-87-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68271

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68271
Availability details for
Quantity
  • Availability
  • Add to Cart