ENA300791826 Enamine

2-(2-aminoethyl)-2,3-dihydro-1lambda6,2-benzothiazole-1,1,3-trione hydrochloride

  • MDL number MFCD16990690
  • Cas Number : 68287-34-3
  • Molecular Weight : 262.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(2-aminoethyl)-1,2-benzisothiazol-3(2H)-one 1,1-dioxide hydrochloride
Inchi Code 1S/C9H10N2O3S.ClH/c10-5-6-11-9(12)7-3-1-2-4-8(7)15(11,13)14;/h1-4H,5-6,10H2;1H
Inchi Key ZPBUZDDTUNLZLK-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-68273

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68273
Availability details for
Quantity
  • Availability
  • Add to Cart