ENA300791827 Enamine

(piperidin-4-yl)methanesulfonamide hydrochloride

  • MDL number MFCD16990691
  • Cas Number : 1251925-40-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-68304

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68304
Availability details for
Quantity
  • Availability
  • Add to Cart