ENA300791940 Enamine

2-(2,3-dihydro-1,4-benzodioxin-6-yl)propanoic acid

  • MDL number MFCD11590415
  • Cas Number : 193885-36-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 81-83
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-67528

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67528
Availability details for
Quantity
  • Availability
  • Add to Cart