ENA300791948 Enamine

1-(5-amino-2-methylphenyl)pyrrolidine-2,5-dione

  • MDL number MFCD09739245
  • Cas Number : 946768-88-3
  • Molecular Weight : 204.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67549

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67549
Availability details for
Quantity
  • Availability
  • Add to Cart