ENA300791954 Enamine

1-(4-{2-[4-(2-aminoethyl)phenoxy]ethyl}piperazin-1-yl)ethan-1-one

  • MDL number MFCD13636612
  • Cas Number : 1094860-51-1
  • Molecular Weight : 291.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67556

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67556
Availability details for
Quantity
  • Availability
  • Add to Cart