ENA300791958 Enamine

2-(pentane-1-sulfonyl)acetic acid

  • MDL number MFCD09940130
  • Cas Number : 107375-91-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67568

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67568
Availability details for
Quantity
  • Availability
  • Add to Cart