ENA300791959 Enamine

2-(3-methoxypropyl)-2,3-dihydro-1H-isoindol-4-amine

  • MDL number MFCD11655099
  • Cas Number : 1099644-34-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-67570
Availability details for
Quantity
  • Availability
  • Add to Cart