ENA300791971 Enamine

2-(2-methoxyethoxy)pyridin-3-amine

  • MDL number MFCD09815653
  • Cas Number : 1016510-18-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67589

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67589
Availability details for
Quantity
  • Availability
  • Add to Cart