ENA300791972 Enamine

3-amino-4-chloro-N-(2-methoxyethyl)benzamide

  • MDL number MFCD09043417
  • Cas Number : 926234-97-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67590

Safety Information

msds https://www.enaminestore.com/catalog/EN300-67590
Availability details for
Quantity
  • Availability
  • Add to Cart