ENA300791974 Enamine

4-bromo-1-(3-methoxypropyl)-1H-pyrazole

  • MDL number MFCD12824379
  • Cas Number : 1183903-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67594

Safety Information

msds https://www.enaminestore.com/catalog/EN300-67594
Availability details for
Quantity
  • Availability
  • Add to Cart