ENA300791977 Enamine

5-methyl-4-(propan-2-yl)-2,3-dihydro-1,3-thiazol-2-one

  • MDL number MFCD12807246
  • Cas Number : 1184200-70-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67600

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67600
Availability details for
Quantity
  • Availability
  • Add to Cart