ENA300791979 Enamine

2-(difluoromethoxy)benzene-1-sulfonamide

  • MDL number MFCD11212973
  • Cas Number : 77798-20-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 133...135
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67254

Safety Information

msds https://www.enaminestore.com/catalog/EN300-67254
Availability details for
Quantity
  • Availability
  • Add to Cart