ENA300791981 Enamine

1-ethyl-2,3-dihydro-1H-indol-6-amine

  • MDL number MFCD11137693
  • Cas Number : 143543-67-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67612

Safety Information

msds https://www.enaminestore.com/catalog/EN300-67612
Availability details for
Quantity
  • Availability
  • Add to Cart