ENA300791994 Enamine

4-[(3-chlorophenyl)methoxy]-2-methylaniline

  • MDL number MFCD09808863
  • Cas Number : 1019110-00-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67632

Safety Information

msds https://www.enaminestore.com/catalog/EN300-67632
Availability details for
Quantity
  • Availability
  • Add to Cart