ENA300791996 Enamine

2-(3-methylbutan-2-yl)-2,3-dihydro-1H-isoindol-4-amine

  • MDL number MFCD11655023
  • Cas Number : 1099671-03-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-67257
Availability details for
Quantity
  • Availability
  • Add to Cart