ENA300792000 Enamine

3-nitro-4-(2,2,2-trifluoroacetamido)benzene-1-sulfonyl chloride

  • MDL number MFCD16990676
  • Cas Number : 1178647-96-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67640

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67640
Availability details for
Quantity
  • Availability
  • Add to Cart