ENA300792002 Enamine

2-(methylamino)thiophene-3-carbonitrile

  • MDL number MFCD05974902
  • Cas Number : 860648-75-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-67649

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67649
Availability details for
Quantity
  • Availability
  • Add to Cart