ENA300792003 Enamine

3-[(4-cyano-2-fluorophenyl)amino]propanamide

  • MDL number MFCD12110080
  • Cas Number : 1157060-76-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67650

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67650
Availability details for
Quantity
  • Availability
  • Add to Cart