ENA300792004 Enamine

N-cyclooctyl-1,3-dioxaindan-5-amine

  • MDL number MFCD01308876
  • Cas Number : 416883-18-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-67260

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67260
Availability details for
Quantity
  • Availability
  • Add to Cart