ENA300792008 Enamine

1-(4-fluorophenyl)-3-(3-hydroxyphenyl)urea

  • MDL number MFCD00091689
  • Cas Number : 199585-00-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-67656

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67656
Availability details for
Quantity
  • Availability
  • Add to Cart