ENA300792013 Enamine

4-(2,3-dihydro-1,4-benzodioxin-6-yl)butan-2-one

  • MDL number MFCD09891024
  • Cas Number : 30830-56-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67662

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67662
Availability details for
Quantity
  • Availability
  • Add to Cart