ENA300792019 Enamine

1-phenylhexan-1-amine

  • MDL number MFCD11184389
  • Cas Number : 92779-83-4
  • Molecular Weight : 177.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-67673

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67673
Availability details for
Quantity
  • Availability
  • Add to Cart