ENA300792023 Enamine

1-(prop-2-yn-1-yl)-1H-pyrazole

  • MDL number MFCD10698153
  • Cas Number : 20306-74-5
  • Molecular Weight : 106.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-67677

Safety Information

signalword Danger
pcodes P210,P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P362,P370+P378,P403+P233,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-67677
pictograms GHS05,GHS07
hstatements H227,H302,H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart