ENA300792026 Enamine

1-(4-chlorophenyl)-3-methylbutan-2-one

  • MDL number MFCD11935569
  • Cas Number : 65248-53-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67794

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67794
Availability details for
Quantity
  • Availability
  • Add to Cart