ENA300792030 Enamine

5-methyl-5-(2-methylphenyl)imidazolidine-2,4-dione

  • MDL number MFCD09731381
  • Cas Number : 88807-84-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169...171
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68154

Safety Information

msds https://www.enaminestore.com/catalog/EN300-68154
Availability details for
Quantity
  • Availability
  • Add to Cart