ENA300792031 Enamine

3-iodo-5-(trifluoromethyl)benzoic acid

  • MDL number MFCD08448293
  • Cas Number : 28186-62-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182...184
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68155

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-68155
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart