ENA300792037 Enamine

6-(2-bromoacetyl)-7-fluoro-1,2,3,4-tetrahydroquinolin-2-one

  • MDL number MFCD12174769
  • Cas Number : 1153886-14-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 224...226
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68167

Safety Information

msds https://www.enaminestore.com/catalog/EN300-68167
Availability details for
Quantity
  • Availability
  • Add to Cart