ENA300792038 Enamine

1-(4-chlorophenyl)-3-methylbutan-2-amine

  • MDL number MFCD11935636
  • Cas Number : 105871-09-8
  • Molecular Weight : 197.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67188

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67188
Availability details for
Quantity
  • Availability
  • Add to Cart