ENA300792039 Enamine

1-(7-methoxy-1-benzofuran-2-yl)methanamine

  • MDL number MFCD12048755
  • Cas Number : 165735-70-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68170

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68170
Availability details for
Quantity
  • Availability
  • Add to Cart