ENA300792041 Enamine

2-(2-chlorophenyl)-2-methylpropan-1-amine

  • MDL number MFCD11043051
  • Cas Number : 1176667-92-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-68172

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68172
Availability details for
Quantity
  • Availability
  • Add to Cart