ENA300792045 Enamine

N-[(2,3-dihydro-1,4-benzodioxin-6-yl)methyl]cyclopentanamine

  • MDL number MFCD01135592
  • Cas Number : 940364-17-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68183

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68183
Availability details for
Quantity
  • Availability
  • Add to Cart