ENA300792046 Enamine

2-(pentan-2-yl)-2,3-dihydro-1H-isoindol-4-amine

  • MDL number MFCD11655051
  • Cas Number : 1099643-86-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68184

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68184
Availability details for
Quantity
  • Availability
  • Add to Cart