ENA300792048 Enamine

1-ethyl-N-(2-methoxyphenyl)piperidin-4-amine

  • MDL number MFCD11141277
  • Cas Number : 1019543-38-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68186

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68186
Availability details for
Quantity
  • Availability
  • Add to Cart